Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ PCR Product Pre-Sequencing Kit

Catalog No. p-7138021
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
100
2000
500 Reactions
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
70-995-1KT 100
70-996-1KT 500 Reactions
70-997-1KT 2000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 70-995-1KT Supplier Applied Biosystems™ Supplier No. 70995.1.KT
Only null left
Add to Cart
Add to Cart

The PCR Product Pre-Sequencing Kit uses a novel, enzymatic clean-up method to pre-treat PCR products prior to sequencing, without any subsequent purification or separation steps.

  • Remove unincorporated primers and nucleotides in about 5 minutes
  • Prepare DNA sequencing samples with one pipetting step
  • Conserve precious PCR products with up to 100% recovery, regardless of amplicon length
  • Minimize spin column or bead clean-up
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -5 to -30°C.
Description PCR Product Pre-Sequencing Kit, 100 rxns, Recombinant, PCR Purification, Uses a Novel, Enzymatic Clean-up Method to Pre-treat PCR Products Prior to Sequencing, without Any Subsequent Purification or Separation Steps
For Use With (Application) PCR Purification
No. of Reactions 100
Product Line ExoSAP-IT
Protein Form Recombinant
Quantity 100 reactions
Research Category Genetic Diseases
Shipping Condition Dry Ice
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.