Learn More
LUNA NANOTECH INC Pcr Product Purification Kit, 50 Preps
Supplier:  LUNA NANOTECH INC NK10550
Encompass_Preferred
PuroSPIN™ PCR Product Purification Kit, 50 Preps
PuroSPIN™ PCR Purification Kit is designed to provide simple and easy-to-use method for purification of PCR DNA products (70 bp − 10 kb) from enzymatic reactions.
The purification technology is based on nucleic acid binding to silica spin-columns, which allows efficient removal of protein, salt and other contaminants.
PuroSPIN™ buffer formulations produce highly purified DNA product for downstream applications, such as sequencing, ligation, transfection, Southern blotting, qPCR, or microarray analysis.
DNA purified by PuroSPIN™ PCR Product Purificaiton Kit shows equal or higher PCR product yield and purity (as determined by the 260/280 and 260/230 ratios) when compared to leading competitors in the field.
Catalog No. 50-256-6949