Learn More
Azzota Corp PCR qPCR plate 96W ABI 50box
Catalog No. 5000886933
Encompass_Preferred
96 WELL PCR/QPCR PLATES 0 1ML NON SKIRTED CLEAR FOR ABI STEPONE PLUS ABI 7500 FAST QUANTSTUDIO 0 1ML Box of 50 pcs
Catalog No. 50-008-86933 Supplier Azzota Corp Supplier No. PCRP96W01NS50
96 WELL PCR/QPCR PLATES 0 1ML NON SKIRTED CLEAR FOR ABI STEPONE PLUS ABI 7500 FAST QUANTSTUDIO 0 1ML Box of 50 pcs