Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Quality Biological Inc Water, PCR Qualified, Pack 10 X 10 mL
SDP

Catalog No. 50983270
Encompass_Preferred

PCR Water is useful for PCR Assays (i.e rRT-PCR, etc.)

DNase, RNase and Protease tested.
Free of contaminants of human, mouse and rat DNA.

Applications: PCR Qualified Water is functionally tested in a PCR reaction. PCR Qualified Water is highly purified water and it is reverse osmosis deionized, diethylpyrocarbonate (DEPC) treated. This product is free of detectable endonucleases, ribonucleases and nicking activity according to the current quality control procedures. Free of detectable contamination of human, mouse and rat DNA.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered and Deionized
DNase, RNase, and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Catalog No. 50-983-270 Supplier Quality Biological Inc Supplier No. 351161671
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PCR Water is useful for PCR Assays (i.e rRT-PCR, etc.)

DNase, RNase and Protease tested.
Free of contaminants of human, mouse and rat DNA.

Applications: PCR Qualified Water is functionally tested in a PCR reaction. PCR Qualified Water is highly purified water and it is reverse osmosis deionized, diethylpyrocarbonate (DEPC) treated. This product is free of detectable endonucleases, ribonucleases and nicking activity according to the current quality control procedures. Free of detectable contamination of human, mouse and rat DNA.

Storage Conditions: 15 °C to 30 °C
Shipping Condition: ambient temperature
Sterile Filtered and Deionized
DNase, RNase, and Protease Tested
Max Shelf Life: 36 months from date of manufacture

Product and/or packaging contains recycled/renewable content. Product and/or packaging is recyclable or reduces waste. Product and/or packaging is safer to human and environmental health.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.