Learn More
Electron Microscopy Sciences Low Temp 96-well PCR Storage Rack, Blue
SDP
Catalog No. 501929489
Encompass_Preferred
The sleek and modern patent pending design of the PCR storage rack offers a unique lid from which the rack slides out and then rotates 180 deg to become a convenient stand. Ergonomic textured finger grips provide a secure hold, reducing the chance of dropping the PCR rack, making transportation easy. To ensure integrity, the translucent polycarbonate lid has a side opening for faster cooling of samples. The base is made from robust ABS construction, with a closed base design to keep samples safe and cold longer. A location ID of numbers and letters allows for organized sample placement for easy and quick sample retrieval. The rack is versatile and can be used on a bench top or for freezer storage. The compact footprint is stackable, which helps save valuable space in the lab. Features a detachable and ambidextrous lid. Holds 0.2 ml tubes, individual or strip. 96 wells, 5.2 mm dia., 8x12 configuration. Store down to -80 deg C.Color: Blue
Catalog No. 50-192-9489 Supplier Electron Microscopy Sciences Supplier No. 61607B
The sleek and modern patent pending design of the PCR storage rack offers a unique lid from which the rack slides out and then rotates 180 deg to become a convenient stand. Ergonomic textured finger grips provide a secure hold, reducing the chance of dropping the PCR rack, making transportation easy. To ensure integrity, the translucent polycarbonate lid has a side opening for faster cooling of samples. The base is made from robust ABS construction, with a closed base design to keep samples safe and cold longer. A location ID of numbers and letters allows for organized sample placement for easy and quick sample retrieval. The rack is versatile and can be used on a bench top or for freezer storage. The compact footprint is stackable, which helps save valuable space in the lab. Features a detachable and ambidextrous lid. Holds 0.2 ml tubes, individual or strip. 96 wells, 5.2 mm dia., 8x12 configuration. Store down to -80 deg C.Color: Blue