Learn More
Chemglass Life Sciences PCR Reaction Strips, 0.2mL, Amplitube, Strip of 8, Natural
SDP
Catalog No. 501532331
Encompass_Preferred
PCR Reaction Strips, 0.2mL, Amplitube, Strip of 8, Natural. Reaction strips include (8) 0.2mL tubes with ultra thin sidewalls and bottoms for more uniform and efficient temperature transfer, therefore reducing PCR reaction time in most 96-well V bottomed thermal cyclers such as MJ Research, Perkin Elmer Hybaid and others. They are more easily handled than single tubes. They will precisely fit standard well spacing and can also be used with 8-channel hand-held pipettors. All strips are molded of polypropylene under the most stringent conditions. Non-attached cap strips are available in dome or flat top designs and ensure a perfect closure during the whole thermal cycle. Cap strips are not included and have to be ordered separately, see listing below. Strips and cap strips are autoclavable. Sold in cases of 125.
Catalog No. 50-153-2331 Supplier Chemglass Life Sciences Supplier No. CLS361001N
PCR Reaction Strips, 0.2mL, Amplitube, Strip of 8, Natural. Reaction strips include (8) 0.2mL tubes with ultra thin sidewalls and bottoms for more uniform and efficient temperature transfer, therefore reducing PCR reaction time in most 96-well V bottomed thermal cyclers such as MJ Research, Perkin Elmer Hybaid and others. They are more easily handled than single tubes. They will precisely fit standard well spacing and can also be used with 8-channel hand-held pipettors. All strips are molded of polypropylene under the most stringent conditions. Non-attached cap strips are available in dome or flat top designs and ensure a perfect closure during the whole thermal cycle. Cap strips are not included and have to be ordered separately, see listing below. Strips and cap strips are autoclavable. Sold in cases of 125.