Learn More
Thomas Scientific PCR Seal 96/8, clear adhesive film, vertically perforated, strong adhesive, 115 x 100 mm, 100/pk
SDP
Catalog No. NC1951667
Encompass
Our PCR Seal is a durable transparent polyester film with a strong adhesive layer. The seal enables a high integrity and efficiently prevents sample evaporation. It is recommended for PCR, qPCR, and other optical applications (e.g. fluorescence or colorimetric measurements) as well as sample storage.
Catalog No. NC1951667 Supplier Thomas Scientific Supplier No. 1149R71
Our PCR Seal is a durable transparent polyester film with a strong adhesive layer. The seal enables a high integrity and efficiently prevents sample evaporation. It is recommended for PCR, qPCR, and other optical applications (e.g. fluorescence or colorimetric measurements) as well as sample storage.