Learn More
Mascon Medical PCR Adhesive Sealing Film, Clear,Optical, Easily simply, Pressure sensitive, 100 pcs/pack
SDP
Catalog No. NC3527258
Encompass_Preferred
PCR Sealing Fim reduces the chance of wel-to-wel contamination and sample evaporation when applied to a microplate.Prevent Loss of PreciousSamples.Use this optically-clear adhesive film to seal the samples into the wells of a microplate.This will reduce the possibiliy ofcross contamination between sample wells and helpensure consistent Real-Time PCR data.For Research Use Only.Not for use in diagnosticprocedures.Easy to Use.Simply peel the backing away from the adhesive fim and apply firmly across the top of your microplate.Stack microplatesand read samples as you would normally except with less chance of sample contamination.Designed to fit on 96-well and384-well plates.FeaturesPressure-sensitiveadhesive provides a tight sealover every wellWill not interfere with sample readingPress-N-Seal:Align properly to the plate,thenpress to sealEasy to apply-will not stick to your glove
Catalog No. NC3527258 Supplier Mascon Medical Supplier No. MM802021B03
PCR Sealing Fim reduces the chance of wel-to-wel contamination and sample evaporation when applied to a microplate.Prevent Loss of PreciousSamples.Use this optically-clear adhesive film to seal the samples into the wells of a microplate.This will reduce the possibiliy ofcross contamination between sample wells and helpensure consistent Real-Time PCR data.For Research Use Only.Not for use in diagnosticprocedures.Easy to Use.Simply peel the backing away from the adhesive fim and apply firmly across the top of your microplate.Stack microplatesand read samples as you would normally except with less chance of sample contamination.Designed to fit on 96-well and384-well plates.FeaturesPressure-sensitiveadhesive provides a tight sealover every wellWill not interfere with sample readingPress-N-Seal:Align properly to the plate,thenpress to sealEasy to apply-will not stick to your glove