Learn More
USA Scientific Inc TempPlate sealing mat for PCR plates, natural
Catalog No. NC9755042
Encompass
TempPlate pressure-fit sealing mat for PCR plates, natural, 5/pack. Non-reactive, medical grade silicone rubber
Catalog No. NC9755042 Supplier USA Scientific Inc Supplier No. 14009605
TempPlate pressure-fit sealing mat for PCR plates, natural, 5/pack. Non-reactive, medical grade silicone rubber