Learn More
Midwest Scientific PR1MA PCR Strip Tubes and Caps, 0.2 mL
SDP
Catalog No. NC1901168
Encompass
PCR Strip Tubes and Caps are evaporation-resistant and provide easy-to-open-and-close lids that prevent sample loss. The uniform wells and ultra-thin walls provide optimal heat transfer and reaction efficiency. Reclosable bags minimize contamination risk and are convenient for opening and closing.
Catalog No. NC1901168 Supplier Midwest Scientific Supplier No. PRPCR28CF
PCR Strip Tubes and Caps are evaporation-resistant and provide easy-to-open-and-close lids that prevent sample loss. The uniform wells and ultra-thin walls provide optimal heat transfer and reaction efficiency. Reclosable bags minimize contamination risk and are convenient for opening and closing.