Learn More
Mascon Medical Mascon Medical™ PCR Tube, 0.2ml, Thin Wall, Transparent, Flat Cap, 500EA/Bag, 2 Bags/Box, 10 Boxes/Case
SDP
Catalog No. NC2458338
Encompass_Preferred
Our Mascon Medical™ high performance 8-Strip 0.2ml Flat Cap PCR Tubes can be used on Reverse Transcription-Polymerase Chain Reaction (RT-PCR) and are made of high-quality, medical grade polypropylene. Our PCR Tubes are DNase/RNase free and are produced in high-grade GMP workshops. They are widely used in genetics, biochemistry, immunology, medicine and other fields, for a variety of processes.Features:•Meets ISO13485 quality management system standard•Can maintain -60Kpa negative pressure test without exploding capSpecifications:•Volume: 0.2ml•Qty. / Case: 10,000•Material: Polypropylene•Sterile: No•Size: Φ7*22.2mm•Temperature Range: -80℃~120℃•Packaging Format: 500 EA/Bag, 2 Bags/Box, 10 Boxes/Case
Catalog No. NC2458338 Supplier Mascon Medical Supplier No. FSPCRTUBE0.2500
Our Mascon Medical™ high performance 8-Strip 0.2ml Flat Cap PCR Tubes can be used on Reverse Transcription-Polymerase Chain Reaction (RT-PCR) and are made of high-quality, medical grade polypropylene. Our PCR Tubes are DNase/RNase free and are produced in high-grade GMP workshops. They are widely used in genetics, biochemistry, immunology, medicine and other fields, for a variety of processes.Features:•Meets ISO13485 quality management system standard•Can maintain -60Kpa negative pressure test without exploding capSpecifications:•Volume: 0.2ml•Qty. / Case: 10,000•Material: Polypropylene•Sterile: No•Size: Φ7*22.2mm•Temperature Range: -80℃~120℃•Packaging Format: 500 EA/Bag, 2 Bags/Box, 10 Boxes/Case