Learn More
USA Scientific Inc 0.2 mL TempAssure PCR tube, attached dome cap, mixed
Catalog No. NC9968701
Encompass
0.2 mL TempAssure PCR tube, attached dome cap, mixed colors. DNase, RNase, DNA, and PCR inhibitor free. 1000/bag.
Catalog No. NC9968701 Supplier USA Scientific Inc Supplier No. 14024308
0.2 mL TempAssure PCR tube, attached dome cap, mixed colors. DNase, RNase, DNA, and PCR inhibitor free. 1000/bag.