Learn More
Research Products International Corp PCR Tube Freezer Box, with Divider Insert, 196 x 0.2ml Tube Capacity, 5 per Case
SDP
Catalog No. 501367677
Encompass
Freezer box with insert designed exclusively for PCR tubes including strip formats and individual tubes. Each box will accommodate up to 196 x 0.2ml tubes in a 14 x 14 array. Box features a one inch shallow depth to provide easy access to tubes. Outer box dimensions are 5 1/4 x 5 1/4 x 1 inches and can be double stacked for storing in a standard freezer rack. Box and insert are suitable for storage at temperatures from 0°C to -80°C. Holds 8 tube strips or 12 tube strips.
Catalog No. 50-136-7677 Supplier Research Products International Corp Supplier No. 181050
Freezer box with insert designed exclusively for PCR tubes including strip formats and individual tubes. Each box will accommodate up to 196 x 0.2ml tubes in a 14 x 14 array. Box features a one inch shallow depth to provide easy access to tubes. Outer box dimensions are 5 1/4 x 5 1/4 x 1 inches and can be double stacked for storing in a standard freezer rack. Box and insert are suitable for storage at temperatures from 0°C to -80°C. Holds 8 tube strips or 12 tube strips.