Learn More
Mascon Medical Mascon Medical™ PCR Tube Rack, 96-Well, 5EA/Bag, 10 Bags/Case
SDP
Catalog No. NC2458343
Encompass_Preferred
Our Mascon Medical™ high performance 96-well PCR Tube Racks can be used to load PCR single tube, 8-strip PCR tube and 96-well PCR plate and protect PCR tube and PCR plate when storing and transporting.Features:•Meets ISO13485 quality management system standard•Reusable•StackableSpecifications:•Volume: 96-well•Qty. / Case: 50•Material: Polypropylene•Color: Transparent cap, yellow base•Sterile: No•Size: 126.3*88*32mm•Temperature Range: -40℃~100℃•Packaging Format: 5 EA/Bag, 10 Bags/Case
Catalog No. NC2458343 Supplier Mascon Medical Supplier No. FSPCRTUBERACK
Our Mascon Medical™ high performance 96-well PCR Tube Racks can be used to load PCR single tube, 8-strip PCR tube and 96-well PCR plate and protect PCR tube and PCR plate when storing and transporting.Features:•Meets ISO13485 quality management system standard•Reusable•StackableSpecifications:•Volume: 96-well•Qty. / Case: 50•Material: Polypropylene•Color: Transparent cap, yellow base•Sterile: No•Size: 126.3*88*32mm•Temperature Range: -40℃~100℃•Packaging Format: 5 EA/Bag, 10 Bags/Case