Learn More
Chemglass Life Sciences PCR Tube Rack, 96-Well (8x12), (5 Racks/1 Lid), 5/Pk, 25/Case
SDP
Catalog No. 501897750
Encompass_Preferred
Fit for 0.2ml tubes and strip tubes and 96 well platesEasily stackable to save spaceTransparent Polycarbonate lid for easy well identificationPolypropylene racks can be autoclaved
Catalog No. 50-189-7750 Supplier Chemglass Life Sciences Supplier No. CGN3617R05
Fit for 0.2ml tubes and strip tubes and 96 well platesEasily stackable to save spaceTransparent Polycarbonate lid for easy well identificationPolypropylene racks can be autoclaved