Learn More
Chemglass Life Sciences PCR Tube Rack, 96-Well (8x12), Individual Lid/Cover (5 Racks/5 Lids), 5/Pk, 25/Case
SDP
Catalog No. 501897749
Encompass_Preferred
Fit for 0.2ml tubes and strip tubes and 96 well platesEasily stackable to save spaceTransparent Polycarbonate lid for easy well identificationPolypropylene racks can be autoclaved
Catalog No. 50-189-7749 Supplier Chemglass Life Sciences Supplier No. CGN3617R01
Fit for 0.2ml tubes and strip tubes and 96 well platesEasily stackable to save spaceTransparent Polycarbonate lid for easy well identificationPolypropylene racks can be autoclaved