Learn More
Research Products International Corp PCR Tube Rack for 0.2ml Micro-Tubes, 3 1/2 x 5 x 1 1/4 Inches, 8 x 12 Array, Assorted Fluorescent Colors, 5 per Case
SDP
Catalog No. 50550279
Encompass
PCR Work-Up can be utilized as a work station or storage rack for a variety of PCR sample processing applications. Rack is formatted for use with 32 x 0.2ml, 24 x 1.5 or 2.0ml, 16 x 0.5ml individual tubes and also 8 or 12 tube strips in an 8 x 12 array. Features alpha numeric indexing for convenient and safe tube identification. Clear, lift off lid offers a non-slip edge for safe stacking of multiple racks and maximizes available bench top space.
Catalog No. 50-550-279 Supplier Research Products International Corp Supplier No. 247677
PCR Work-Up can be utilized as a work station or storage rack for a variety of PCR sample processing applications. Rack is formatted for use with 32 x 0.2ml, 24 x 1.5 or 2.0ml, 16 x 0.5ml individual tubes and also 8 or 12 tube strips in an 8 x 12 array. Features alpha numeric indexing for convenient and safe tube identification. Clear, lift off lid offers a non-slip edge for safe stacking of multiple racks and maximizes available bench top space.