Learn More
Research Products International Corp PCR Tube Rack for 0.2ml Micro-Tubes, 8 x 12 Array, Assorted Standard Colors, 5 per Case
SDP
Catalog No. 50476667
Encompass
Perfect for pre or post PCR sampling, rack holds 96 x 0.2ml individual PCR tubes or 8 x 12 tube strips. Manufactured from heavy duty polypropylene with a convenient hinged lid that can be removed. Racks feature imprinted alpha/numeric grid identification for convenient tube location. Easy stacking and lid clicks into place.
Catalog No. 50-476-667 Supplier Research Products International Corp Supplier No. 353110
Perfect for pre or post PCR sampling, rack holds 96 x 0.2ml individual PCR tubes or 8 x 12 tube strips. Manufactured from heavy duty polypropylene with a convenient hinged lid that can be removed. Racks feature imprinted alpha/numeric grid identification for convenient tube location. Easy stacking and lid clicks into place.