Learn More
USA Scientific Inc Stackable PCR tube rack, mixed colors
Catalog No. NC9060996
Encompass
Stackable polypropylene PCR tube rack, mixed colors, 5 1/2 in L x 4 1/8 in W x 1 9/16 in H, 5/pack
Catalog No. NC9060996 Supplier USA Scientific Inc Supplier No. 23009708
Stackable polypropylene PCR tube rack, mixed colors, 5 1/2 in L x 4 1/8 in W x 1 9/16 in H, 5/pack