Learn More
Axygen™ PCR Tube Storage Racks
GSA_VAGREENER_CHOICE
Catalog No. 14222396 Shop All Axygen Products
Change view
Click to view available options
Color:
Fluorescent Blue, Green, Red, Violet, Yellow
Fluorescent Yellow
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
14-222-396 Fluorescent Blue, Green, Red, Violet, Yellow
14-222-397 Fluorescent Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-222-396 Supplier Axygen™ Supplier No. R96PCRFSP
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Add to Cart
Constructed of polypropylene. Corning™ Axygen™ PCR Tube Storage Racks hold 96 0.2mL PCR tubes. Supplied with lids.
Polypropylene rack for PCR Tubes
  • Holds 96 0.2mL PCR tubes
  • Available in several colors
  • Supplied with lids
  • Packaged five per pack and five packs per case. Spectrum pack includes five racks of each fluorescent color
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
Color Fluorescent Blue, Green, Red, Violet, Yellow
Holds Microtubes (0.2 to 0.5 mL)
Material Polycarbonate
Positions 96
Quantity 25/Cs.
Includes Clear Polycarbonate Cover
Product Type Microtube Rack
WARNING: Cancer - www.P65Warnings.ca.gov