Learn More
Research Products International Corp PCR Unirack, Blue, 10 per Case
SDP
Catalog No. 50212887
Encompass
Most versatile micro-tube storage system available. Ideal for use as a workstation or storage rack. Holds a combination of up to sixty 0.2 ml and 0.5 ml PCR tubes on one side or 80 x 1.5 ml micro-tubes on reverse side. Removable handles facilitate carrying and stacking. Optional clear PVC lid locks onto rack for secure protection.
Catalog No. 50-212-887 Supplier Research Products International Corp Supplier No. 353082B
Most versatile micro-tube storage system available. Ideal for use as a workstation or storage rack. Holds a combination of up to sixty 0.2 ml and 0.5 ml PCR tubes on one side or 80 x 1.5 ml micro-tubes on reverse side. Removable handles facilitate carrying and stacking. Optional clear PVC lid locks onto rack for secure protection.