Learn More
TWD Scientific LLC Pcr Tube & Cap, Twdbiox, 0.2Ml, Polypropylene, Thin Wall Tube, Frosted Flat Top Cap, 10000/Case.
SDP
Catalog No. 502285002 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 0.2Ml Clear Polypropylene Thin Wall Pcr Tubes With Attached Frosted Flat Top Caps. 1000 Tubes/Pack, 10 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5002 Supplier TWD Scientific LLC Supplier No. BX603220
BioX 0.2Ml Clear Polypropylene Thin Wall Pcr Tubes With Attached Frosted Flat Top Caps. 1000 Tubes/Pack, 10 Packs/Case, Price/Case