Learn More
TWD Scientific LLC Pcr Tube & Cap, Twdbiox, 0.2Ml, Polypropylene, Thin Wall Tube, Clear Flat Top Cap, Strips Of 8, 1200/Case.
SDP
Catalog No. 502285007 Shop All TWD Scientific LLC Products
Encompass_Preferred
BioX 0.2Ml Clear Polypropylene Thin Wall Pcr Tubes With Clear Flat Top Caps. Strong Link Between Tubes With Cap Hinge At 45º Angle. Tubes & Caps Come In Strips Of 8. 120 Strips/Pack, 10 Packs/Case, Price/Case
This item is not returnable. View return policy
Catalog No. 50-228-5007 Supplier TWD Scientific LLC Supplier No. BX603247
BioX 0.2Ml Clear Polypropylene Thin Wall Pcr Tubes With Clear Flat Top Caps. Strong Link Between Tubes With Cap Hinge At 45º Angle. Tubes & Caps Come In Strips Of 8. 120 Strips/Pack, 10 Packs/Case, Price/Case