Learn More
Research Products International Corp PCR Unirack, Orange, 10 per Case
SDP
Catalog No. 501978384
Encompass
Most versatile micro-tube storage system available. Ideal for use as a workstation or storage rack. Holds a combination of up to sixty 0.2 ml and 0.5 ml PCR tubes on one side or 80 x 1.5 ml micro-tubes on reverse side. Removable handles facilitate carrying and stacking. Optional clear PVC lid locks onto rack for secure protection.
Catalog No. 50-197-8384 Supplier Research Products International Corp Supplier No. 353082O
Most versatile micro-tube storage system available. Ideal for use as a workstation or storage rack. Holds a combination of up to sixty 0.2 ml and 0.5 ml PCR tubes on one side or 80 x 1.5 ml micro-tubes on reverse side. Removable handles facilitate carrying and stacking. Optional clear PVC lid locks onto rack for secure protection.