Learn More
OHAUS PCR Vortex Genie Kit
Catalog No. 01804382 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Vortex Attachment
Catalog No. Product Type
01-804-382 Vortex Attachment
1 options
Catalog No. 01-804-382 Supplier OHAUS Supplier No. 83041424
These longer stiffer bumpers replace the existing bumpers.
  • It is compatible with the Digital MicroPlate Genie Pulse and Digital Multi-MicroPlate Genie Pulse Mixers to accommodate the taller microplates and PCR Plates.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Vortex Attachment
Model MicroPlate Genie, Digital MicroPlate Genie Pulse Multi-MicroPlate Genie, and Digital Multi-MicroPlate Genie Pulse Mixers