This PCR workstation provides a sterile, contaminant-free environment for non-hazardous applications by utilizing vertical laminar airflow and HEPA filtration to maintain ISO Class 5 air cleanliness. The unit features durable, epoxy-coated steel construction and UV-absorbing polycarbonate panels for 360 degree visibility, with an integrated germicidal UV lamp and timer for efficient decontamination.
- Main HEPA filter with 99.99% efficiency for 0.3 μm particulates
- Vertical laminar airflow with a nominal velocity of up to 0.45 m/s (90 fpm)
- Germicidal 15 watt UV lamp with safety interlock and 0 to 60 minute timer
- Ergonomic angled front sash for improved user comfort and access
- Energy-saving LED lighting
- Dual rear wall pass-through ports for routing instrument cables
- Includes a rear-mounted decontamination shelf for maximum UV exposure