Learn More
Grainger Air Science Purair PCR Workstation, Polycarbonate, 48 in., 115VAC
Catalog No. 502775747
Encompass_Preferred
The Purair PCR workstation provides a sterile work zone for PCR amplification and other non-hazardous applications sensitive to contamination. Featuring vertical laminar airflow with HEPA filtration, this unit ensures high air cleanliness standards for precise laboratory processes.
  • Provides a sterile work zone for aseptic techniques
  • Meets and exceeds ISO Class 5 air cleanliness
  • Constructed from UV-absorbing polycarbonate for 360 degree visibility
  • Equipped with a germicidal UV lamp for safe decontamination
  • Incorporates energy-saving LED lighting
  • Includes a blower, work surface, and base
  • Designed for desktop use with 115VAC, 60Hz operation
  • Features HEPA filtration with 99.99% efficiency at 0.3 μm
Catalog No. 50-277-5747 Supplier Grainger Supplier No. 18AX43
The Purair PCR workstation provides a sterile work zone for PCR amplification and other non-hazardous applications sensitive to contamination. Featuring vertical laminar airflow with HEPA filtration, this unit ensures high air cleanliness standards for precise laboratory processes.
  • Provides a sterile work zone for aseptic techniques
  • Meets and exceeds ISO Class 5 air cleanliness
  • Constructed from UV-absorbing polycarbonate for 360 degree visibility
  • Equipped with a germicidal UV lamp for safe decontamination
  • Incorporates energy-saving LED lighting
  • Includes a blower, work surface, and base
  • Designed for desktop use with 115VAC, 60Hz operation
  • Features HEPA filtration with 99.99% efficiency at 0.3 μm