Learn More
Grainger Air Science Purair PCR Workstation, Polycarbonate, 24 in., HEPA
Catalog No. 502777704
Encompass_Preferred
The Purair® PCR workstation provides a protected, vertical laminar flow environment designed specifically for sensitive PCR amplification procedures. By utilizing HEPA filtration and a germicidal UV decontamination cycle, the cabinet maintains an ISO Class 5 air cleanliness level. It is constructed from UV-absorbing polycarbonate to ensure 360 degree visibility while protecting the user from germicidal UV emission. This unit is intended for non-hazardous research protocols where biological or biohazard byproducts are not generated.
  • Vertical laminar airflow with HEPA filtration achieving 99.97% efficiency at 0.3 μm.
  • Equipped with a 253.7 nm germicidal UV lamp and timer for decontamination.
  • Magnetic door latch and safety interlock.
  • High-performance EC fan with solid-state speed control.
  • Convenient decontamination shelf mounted on the rear wall.
Catalog No. 50-277-7704 Supplier Grainger Supplier No. 18AX52
The Purair® PCR workstation provides a protected, vertical laminar flow environment designed specifically for sensitive PCR amplification procedures. By utilizing HEPA filtration and a germicidal UV decontamination cycle, the cabinet maintains an ISO Class 5 air cleanliness level. It is constructed from UV-absorbing polycarbonate to ensure 360 degree visibility while protecting the user from germicidal UV emission. This unit is intended for non-hazardous research protocols where biological or biohazard byproducts are not generated.
  • Vertical laminar airflow with HEPA filtration achieving 99.97% efficiency at 0.3 μm.
  • Equipped with a 253.7 nm germicidal UV lamp and timer for decontamination.
  • Magnetic door latch and safety interlock.
  • High-performance EC fan with solid-state speed control.
  • Convenient decontamination shelf mounted on the rear wall.