Learn More
MBF Bioscience PCR Workstations — Dead Air Boxes

Description
Includes
Acid-resistant Formica™ work surface, tempered safety-glass fascia, duplex electrical outlet mounted in ceiling, two fluorescent lamps mounted in stainless-steel ceiling, single or dual UV germicidal lamp with 12-hour countdown timer with time-hold position and single or double door closures
Three sizes offer plenty of workspace for reaction supplies and thermal cycler.
- UV irradiation of working area prior to use blocks replication of contaminating DNA sequences by causing adjacent pyrimidines to undergo dimerization
- Protects against cross- or airborne contamination by limiting exposure of experimental setup to open lab environment
- Dual UV bulbs deliver twice the UV intensity compared to single bulbs, helping irradiate areas that might otherwise be inaccessible; useful when workstation is used to decontaminate apparatus and reagents
- Access doors close to prevent contamination, then slide into storage compartment in base during use
- Stainless-steel ceiling increases UV dose at workstation surface; results in a 10% increase in UV dose with single bulb and 20% with dual UV bulb
- Hinged-glass fascia allows easy access to interior workspace for cleaning and placement of large instruments
- Dual fluorescent lamps ensure excellent workspace visibility
- 254mm UV lamp with convenient switch for timed decontamination of workspace
Order Info
110V.
Certifications
CE-marked.
Specifications
Specifications
| Product Type | PCR Enclosure |
| Lighting Type | Fluorescent, UV |
| Certifications/Compliance | CE |
| Depth (English) | 24 in. |
| Depth (Metric) | 60.9 cm |
| For Use With (Application) | Protect against contamination in sensitive PCR amplification reactions |
| Height (English) | 24 in. |
| Height (Metric) | 60.9 cm |
| Includes | 1 UV, 2 fluorescent bulbs |
| Voltage | 110 V |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.