Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CBS Scientific PCR Workstations — Dead Air Boxes
SCI_ED

Catalog No. S05781
Change view
Click to view available options
Includes:
1 UV, 2 fluorescent bulbs
2 UV, 2 fluorescent bulbs
Width (Metric):
76 cm
76.2 cm
91.4 cm
122 cm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Width (Metric)
S05781 2 UV, 2 fluorescent bulbs 76.2 cm
S05780 1 UV, 2 fluorescent bulbs 76 cm
S05783 2 UV, 2 fluorescent bulbs 91.4 cm
S05782 1 UV, 2 fluorescent bulbs 91.4 cm
S05784 1 UV, 2 fluorescent bulbs 122 cm
S05785 2 UV, 2 fluorescent bulbs 122 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S05781 Supplier CBS Scientific Supplier No. P030202
Add to Cart
Add to Cart

Includes

Acid-resistant Formica™ work surface, tempered safety-glass fascia, duplex electrical outlet mounted in ceiling, two fluorescent lamps mounted in stainless-steel ceiling, single or dual UV germicidal lamp with 12-hour countdown timer with time-hold position and single or double door closures

Protect against contamination in sensitive PCR amplification reactions.

Three sizes offer plenty of workspace for reaction supplies and thermal cycler.

  • UV irradiation of working area prior to use blocks replication of contaminating DNA sequences by causing adjacent pyrimidines to undergo dimerization
  • Protects against cross- or airborne contamination by limiting exposure of experimental setup to open lab environment

  • Dual UV bulbs deliver twice the UV intensity compared to single bulbs, helping irradiate areas that might otherwise be inaccessible; useful when workstation is used to decontaminate apparatus and reagents
  • Access doors close to prevent contamination, then slide into storage compartment in base during use
  • Stainless-steel ceiling increases UV dose at workstation surface; results in a 10% increase in UV dose with single bulb and 20% with dual UV bulb
  • Hinged-glass fascia allows easy access to interior workspace for cleaning and placement of large instruments
  • Dual fluorescent lamps ensure excellent workspace visibility
  • 254mm UV lamp with convenient switch for timed decontamination of workspace

Order Info

110V.

Certifications

CE-marked.

Specifications

Product Type PCR Enclosure
Lighting Type Fluorescent, UV
Certifications/Compliance CE
Depth (English) 24 in.
Depth (Metric) 60.9 cm
For Use With (Application) Protect against contamination in sensitive PCR amplification reactions
Height (English) 24 in.
Height (Metric) 60.9 cm
Includes 2 UV, 2 fluorescent bulbs
Voltage 110 V
Width (English) 30 in.
Width (Metric) 76.2 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.