Learn More
Research Products International Corp PCRack with Asstd Lid 4 pk
SDP
Catalog No. 502388451
Encompass
This convenient space-saving rack was designed especially for storing and working with PCR samples. The PCRack will accept all models of 0.2 ml tubes, strips of 8 or 12 tubes and 96-well PCR plates . Stackable racks are made of polypropylene and can be horizontally attached to each other in order to build up any configuration. With the cover on, they are easily stackable one on top of another. Thanks to these special features, efficiency is highly improved allowing you to carry a multitude of tubes and/or strips at the same time. Available in 6 colors or assorted pack (blue, green, pink, and yellow).
Stackable
Improve efficiency of PCR storage
Available in 6 colors or assorted pack
Catalog No. 50-238-8451 Supplier Research Products International Corp Supplier No. 400706A
This convenient space-saving rack was designed especially for storing and working with PCR samples. The PCRack will accept all models of 0.2 ml tubes, strips of 8 or 12 tubes and 96-well PCR plates . Stackable racks are made of polypropylene and can be horizontally attached to each other in order to build up any configuration. With the cover on, they are easily stackable one on top of another. Thanks to these special features, efficiency is highly improved allowing you to carry a multitude of tubes and/or strips at the same time. Available in 6 colors or assorted pack (blue, green, pink, and yellow).
Stackable
Improve efficiency of PCR storage
Available in 6 colors or assorted pack