Learn More
Cayman Chemical Akuammine, Purity: ≥98%, CAS: 3512-87-6, 10 mg
SDP
Catalog No. 502910378
Encompass_Preferred
An indole alkaloid with analgesic activity; selectively binds the μ- and κ-opioid receptors over the δ-opioid receptor (Kis = 0.3, 1.68, and 10.4 µM for the human receptors, respectively); inhibits forskolin-induced cAMP production in HEK293 cells expressing human μ- or κ-opioid receptors (IC50s = 2.6 and 0.073 µM, respectively); increases the latency to withdrawal in the tail-flick or hot plate test in mice at 60 mg/kg
Catalog No. 50-291-0378 Supplier Cayman Chemical Supplier No. 3514910 MG
An indole alkaloid with analgesic activity; selectively binds the μ- and κ-opioid receptors over the δ-opioid receptor (Kis = 0.3, 1.68, and 10.4 µM for the human receptors, respectively); inhibits forskolin-induced cAMP production in HEK293 cells expressing human μ- or κ-opioid receptors (IC50s = 2.6 and 0.073 µM, respectively); increases the latency to withdrawal in the tail-flick or hot plate test in mice at 60 mg/kg