Learn More
Advanced Instruments Inc Artel PCS Pipette Calibration System, High-Precision, Portable, ISO Compliant, Units: 1 Each
SDP
Catalog No. NC3779060
Encompass
The Advanced Instruments Artel PCS Pipette Calibration System is a high-precision, portable solution for volume verification and calibration of single-channel pipettes. It ensures data integrity and compliance with regulatory standards through its dual-dye ratiometric photometric measurement method. Designed for seamless integration with ArtelWare™ Software, it enhances workflow efficiency by offering features like calibration scheduling and real-time documentation, making it a reliable choice for laboratories aiming to streamline pipette management.
Catalog No. NC3779060 Supplier Advanced Instruments Inc Supplier No. PCS512NC
Add to Cart
Add to Cart
Request a Quote
The Advanced Instruments Artel PCS Pipette Calibration System is a high-precision, portable solution for volume verification and calibration of single-channel pipettes. It ensures data integrity and compliance with regulatory standards through its dual-dye ratiometric photometric measurement method. Designed for seamless integration with ArtelWare™ Software, it enhances workflow efficiency by offering features like calibration scheduling and real-time documentation, making it a reliable choice for laboratories aiming to streamline pipette management.