Learn More
Accuflow Systems PCS Pump Tubing
Supplier:  Accuflow Systems ONESILP00630188
Encompass_Preferred
Purity One™ OneSil Pump biopharmaceutical grade silicone tubing has been formulated for demanding pumping applications in biopharmaceutical processing. This low-hysteresis pump grade silicone tubing is moldable for single-use assemblies, has higher pressure ratings and better kink resistance than traditional 50D silicone tubing, and has a large temperature range, making it an excelling choice for pumping, sampling, filtration, sterile filling and transfer, and buffer and media prep. Full validation guide and USP <665> Extractables testing available.
Catalog No. 50-299-5726