Learn More
Advanced Instruments Inc PCS STD REAGENT KIT
SDP
Catalog No. NC9669282
Encompass
Standard Reagent Kit, 12 Blank Vials, 1 zero Calibrator, aliquot containers, transfer pipettes, sample solution for ranges 1-4
Catalog No. NC9669282 Supplier Advanced Instruments Inc Supplier No. PCS200
Standard Reagent Kit, 12 Blank Vials, 1 zero Calibrator, aliquot containers, transfer pipettes, sample solution for ranges 1-4