Learn More
Aviva Systems Biology Corp PCSK2 Antibody - middle region (ARP59552_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59552_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PCSK2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PCSK2. Alternate names: NEC2, PC2, SPC2, NEC 2, NEC-2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-738