Learn More
ACROBiosystems ACROBiosystems Cynomolgus PCSK9 Protein, His Tag
Supplier:  ACROBiosystems PC9C522350UG
Encompass_Preferred
Recombinant Protein;50UG;Cynomolgus PCSK9, His Tag (PC9-C5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # G7NVZ1-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 73.2 kDa. The protein migrates as 17 kDa and 65 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human LDL R, Strep Tag (Cat. No. LDR-H5281) at 2 μg/mL (100 μL/well) can bind Cynomolgus PCSK9, His Tag (Cat. No. PC9-C5223) with a linear range of 4-31 ng/mL (QC tested).
Catalog No. 50-195-7055