Learn More
Medchemexpress LLC PCSK9 Human Biotin 100ug
SDP
Supplier:  Medchemexpress LLC HYP72405100UG
Encompass_Preferred
The PCSK9 protein is an important regulator of plasma cholesterol homeostasis affecting LDL receptor family members such as LDLR and VLDLR It promotes their intracellular degradation and enhances hepatic LDLR degradation through non-proteolytic mechanisms PCSK9 Protein Human (Biotinylated V474I G670E HEK293 Avi) is the recombinant human-derived PCSK9 protein expressed by HEK293 with C-Avi labeled tag and V474I G670E mutation
Catalog No. 50-001-82705