Learn More
Medchemexpress LLC PCSK9 Human Biotin 20ug
SDP
Catalog No. 5000181960
Encompass_Preferred
The PCSK9 protein is an important regulator of plasma cholesterol homeostasis affecting LDL receptor family members such as LDLR and VLDLR It promotes their intracellular degradation and enhances hepatic LDLR degradation through non-proteolytic mechanisms PCSK9 Protein Human (Biotinylated V474I G670E HEK293 Avi) is the recombinant human-derived PCSK9 protein expressed by HEK293 with C-Avi labeled tag and V474I G670E mutation
Catalog No. 50-001-81960 Supplier Medchemexpress LLC Supplier No. HYP7240520UG
The PCSK9 protein is an important regulator of plasma cholesterol homeostasis affecting LDL receptor family members such as LDLR and VLDLR It promotes their intracellular degradation and enhances hepatic LDLR degradation through non-proteolytic mechanisms PCSK9 Protein Human (Biotinylated V474I G670E HEK293 Avi) is the recombinant human-derived PCSK9 protein expressed by HEK293 with C-Avi labeled tag and V474I G670E mutation