Learn More
Medchemexpress LLC PCSK9 Human Biotin 5ug | 5ug
SDP
Catalog No. 5000194865
Encompass_Preferred
The PCSK9 protein is an important regulator of plasma cholesterol homeostasis affecting LDL receptor family members such as LDLR and VLDLR It promotes their intracellular degradation and enhances hepatic LDLR degradation through non-proteolytic mechanisms PCSK9 Protein Human (Biotinylated HEK293 D374Y His-Avi) is the recombinant human-derived PCSK9 protein expressed by HEK293 with C-Avi C-His labeled tag and D374Y mutation
Catalog No. 50-001-94865 Supplier Medchemexpress LLC Supplier No. HYP788245UG
The PCSK9 protein is an important regulator of plasma cholesterol homeostasis affecting LDL receptor family members such as LDLR and VLDLR It promotes their intracellular degradation and enhances hepatic LDLR degradation through non-proteolytic mechanisms PCSK9 Protein Human (Biotinylated HEK293 D374Y His-Avi) is the recombinant human-derived PCSK9 protein expressed by HEK293 with C-Avi C-His labeled tag and D374Y mutation