Learn More
Sino Biological PCSK9 / NARC1 Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 10594MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PCSK9 (rhPCSK9; Catalog#10594-H08H; Met1-Gln692; AAV67948.1). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8521