Learn More
Sino Biological PCSK9 / NARC1 Antibody, Rabbit PAb 100uL
Supplier:  Sino Biological 10594RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PCSK9 / NARC1 (rh PCSK9; Catalog#10594-H08H; AAV67948.1; Met1-Gln692). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-224-8524