Learn More
ACROBiosystems Recombinant Protein;Human PCSK9 Protein, Fc Tag;HEK293;1MG;PC9,PCSK9,PCSK-9,FH3,HCHOLA3,LDLCQ1,NARC-1,NARC1,PC9
Catalog No. 501657536
Encompass_Preferred
Recombinant Protein;Human PCSK9 Protein, Fc Tag;1MG;Human PCSK9, Fc Tag (PC9-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # Q8NBP7-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 97.5 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VLDL R, His Tag (Cat. No. VLR-H5227) at 5 μg/mL (100 μL/well) can bind Human PCSK9, Fc Tag (Cat. No. PC9-H5256) with a linear range of 1-10 ng/mL (QC tested).;Immobilized Human PCSK9, Fc Tag (Cat. No. PC9-H5256) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LDL R, His Tag, Avi Tag (Cat. No. LDR-H82E7) with a linear range of 0.04-0.625 μg/mL (Routinely tested).
Catalog No. 50-165-7536 Supplier ACROBiosystems Supplier No. PC9H52561MG
Recombinant Protein;Human PCSK9 Protein, Fc Tag;1MG;Human PCSK9, Fc Tag (PC9-H5256) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # Q8NBP7-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 97.5 kDa. The protein migrates as 100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human VLDL R, His Tag (Cat. No. VLR-H5227) at 5 μg/mL (100 μL/well) can bind Human PCSK9, Fc Tag (Cat. No. PC9-H5256) with a linear range of 1-10 ng/mL (QC tested).;Immobilized Human PCSK9, Fc Tag (Cat. No. PC9-H5256) at 5 μg/mL (100 μL/well) can bind Biotinylated Human LDL R, His Tag, Avi Tag (Cat. No. LDR-H82E7) with a linear range of 0.04-0.625 μg/mL (Routinely tested).