Learn More
ACROBiosystems Recombinant Protein;Mouse PCSK9 Protein;HEK293;1MG;PC9,PCSK9,PCSK-9,FH3,HCHOLA3,LDLCQ1,NARC-1,NARC1,PC9
Supplier:  ACROBiosystems PC9M52281MG
Encompass_Preferred
Recombinant Protein;Mouse PCSK9 Protein;1MG;Mouse PCSK9, His Tag (PC9-M5228) is expressed from human 293 cells (HEK293). It contains AA Gln 35 - Gln 694 (Accession # AAH38085.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 72 kDa. The protein migrates as 19 kDa and 57-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation and proteolytic digestion.;Immobilized Human LDL R, Strep Tag (Cat. No. LDR-H5281) at 10 μg/mL (100 μl/well) can bind Mouse PCSK9, His Tag (Cat. No. PC9-M5228 ) with a linear range of 2-30 ng/mL (QC tested).
Catalog No. 50-165-7811