Learn More
ACROBiosystems Recombinant Protein;Cynomolgus PCSK9 Protein, His Tag;HEK293;1MG;PC9,PCSK9,PCSK-9,FH3,HCHOLA3,LDLCQ1,NARC-1,NARC1,PC9
Catalog No. 501657437
Encompass_Preferred
Recombinant Protein;Cynomolgus PCSK9 Protein, His Tag;1MG;Cynomolgus PCSK9, His Tag (PC9-C5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # G7NVZ1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 73.2 kDa. The protein migrates as 17 kDa and 65 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human LDL R, Strep Tag (Cat. No. LDR-H5281) at 2 μg/mL (100 μL/well) can bind Cynomolgus PCSK9, His Tag (Cat. No. PC9-C5223) with a linear range of 4-31 ng/mL (QC tested).
Catalog No. 50-165-7437 Supplier ACROBiosystems Supplier No. PC9C52231MG
Recombinant Protein;Cynomolgus PCSK9 Protein, His Tag;1MG;Cynomolgus PCSK9, His Tag (PC9-C5223) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # G7NVZ1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 73.2 kDa. The protein migrates as 17 kDa and 65 kDa under reducing (R) condition (SDS-PAGE).;Immobilized Human LDL R, Strep Tag (Cat. No. LDR-H5281) at 2 μg/mL (100 μL/well) can bind Cynomolgus PCSK9, His Tag (Cat. No. PC9-C5223) with a linear range of 4-31 ng/mL (QC tested).