Learn More
ACROBiosystems Recombinant Protein;Rat PCSK9 Protein;HEK293;50UG;PC9,PCSK9,PCSK-9,FH3,HCHOLA3,LDLCQ1,NARC-1,NARC1,PC9
Supplier: ACROBiosystems PC9R52H450UG
Recombinant Protein;Rat PCSK9 Protein;50UG;Rat PCSK9, His Tag (PC9-R52H4) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 691 (Accession # P59996-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 73.2 kDa. The protein migrates as 20 kDa and 63-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LDL R, Strep Tag (Cat. No. LDR-H5281) at 2 μg/mL (100 μL/well) can bind Rat PCSK9, His Tag (Cat. No. PC9-R52H4) with a linear range of 8-63 ng/mL (QC tested).