Learn More
ACROBiosystems Recombinant Protein;Hamster PCSK9 Protein;HEK293;50UG;PC9,PCSK9,PCSK-9,FH3,HCHOLA3,LDLCQ1,NARC-1,NARC1,PC9
Catalog No. 501659086
Encompass_Preferred
Recombinant Protein;Hamster PCSK9 Protein;50UG;Hamster PCSK9, His Tag (PC9-H52E4) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Ser 691 (Accession # G3GTK5).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 73 kDa. The protein migrates as 18 kDa and 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation and proteolytic digestion.;Immobilized Human LDL R (High Purity) Protein, Strep Tag (Cat. No. LDR-H5281) at 5 μg/mL (100 μl/well) can bind Hamster PCSK9 Protein, His Tag (Cat. No. PC9-H52E4) with a linear range of 6-50 ng/mL (QC tested).
Catalog No. 50-165-9086 Supplier ACROBiosystems Supplier No. PC9H52E450UG
Recombinant Protein;Hamster PCSK9 Protein;50UG;Hamster PCSK9, His Tag (PC9-H52E4) is expressed from human 293 cells (HEK293). It contains AA Gln 30 - Ser 691 (Accession # G3GTK5).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 73 kDa. The protein migrates as 18 kDa and 65 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation and proteolytic digestion.;Immobilized Human LDL R (High Purity) Protein, Strep Tag (Cat. No. LDR-H5281) at 5 μg/mL (100 μl/well) can bind Hamster PCSK9 Protein, His Tag (Cat. No. PC9-H52E4) with a linear range of 6-50 ng/mL (QC tested).