Learn More
ACROBiosystems Recombinant Protein;Rhesus macaque PCSK9 Protein;HEK293;50UG;PC9,PCSK9,PCSK-9,FH3,HCHOLA3,LDLCQ1,NARC-1,NARC1,PC9
Catalog No. 501658566
Encompass_Preferred
Recombinant Protein;Rhesus macaque PCSK9 Protein;50UG;Rhesus macaque PCSK9, His Tag (PC9-C52H2) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # A8T666).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 73.2 kDa. The protein migrates as 21 kDa and 64 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LDL R, Strep Tag (Cat. No. LDR-H5281) at 2 μg/mL (100 μL/well) can bind Rhesus macaque PCSK9, His Tag (Cat. No. PC9-C52H2) with a linear range of 1-15.6 ng/mL (QC tested).
Catalog No. 50-165-8566 Supplier ACROBiosystems Supplier No. PC9C52H250UG
Recombinant Protein;Rhesus macaque PCSK9 Protein;50UG;Rhesus macaque PCSK9, His Tag (PC9-C52H2) is expressed from human 293 cells (HEK293). It contains AA Gln 31 - Gln 692 (Accession # A8T666).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 73.2 kDa. The protein migrates as 21 kDa and 64 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human LDL R, Strep Tag (Cat. No. LDR-H5281) at 2 μg/mL (100 μL/well) can bind Rhesus macaque PCSK9, His Tag (Cat. No. PC9-C52H2) with a linear range of 1-15.6 ng/mL (QC tested).