Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human PCSK9, Avi Tag (Avitag™);HEK293;25UG;PC9,PCSK9,PCSK-9,FH3,HCHOLA3,LDLCQ1,NARC-1,NARC1,PC9
Catalog No. 501659154
Encompass_Preferred
Recombinant Protein;Biotinylated Human PCSK9, Avi Tag (Avitag™);25UG;MABSol Biotinylated Human PCSK9 (PC9-H82E7) is expressed from human HEK293 cells. It contains AA Gln 31 - Gln 692 (Accession # AAI66619).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 72 kDa. The protein migrates as 20 kDa and 66 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation and proteolytic digestion.;Immobilized Human LDL R, His Tag (Cat. No. LDR-H5224) at 10 g/mL (100 L/well) can bind Biotinylated Human PCSK9, His Tag (Cat. No. PC9-H82E7) with a linear range of 2-31 ng/mL (QC tested).;Immobilized Biotinylated Human PCSK9, His Tag (Cat. No. PC9-H82E7) can bind anti-Human PCSK9 Mab, Human IgG1 with a linear range of 0.05-1.5 ng/mL. The biotinylated PCSK9 is coated at 1 ug/mL (100 uL/well) on a streptavidin coated plates (Routinely tested).
Catalog No. 50-165-9154 Supplier ACROBiosystems Supplier No. PC9H82E725UG
Recombinant Protein;Biotinylated Human PCSK9, Avi Tag (Avitag™);25UG;MABSol Biotinylated Human PCSK9 (PC9-H82E7) is expressed from human HEK293 cells. It contains AA Gln 31 - Gln 692 (Accession # AAI66619).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 72 kDa. The protein migrates as 20 kDa and 66 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation and proteolytic digestion.;Immobilized Human LDL R, His Tag (Cat. No. LDR-H5224) at 10 g/mL (100 L/well) can bind Biotinylated Human PCSK9, His Tag (Cat. No. PC9-H82E7) with a linear range of 2-31 ng/mL (QC tested).;Immobilized Biotinylated Human PCSK9, His Tag (Cat. No. PC9-H82E7) can bind anti-Human PCSK9 Mab, Human IgG1 with a linear range of 0.05-1.5 ng/mL. The biotinylated PCSK9 is coated at 1 ug/mL (100 uL/well) on a streptavidin coated plates (Routinely tested).