Learn More
ACROBiosystems Recombinant Protein;Biotinylated Mouse PCSK9, Avi Tag (Avitag™);HEK293;25UG;PC9,PCSK9,PCSK-9,FH3,HCHOLA3,LDLCQ1,NARC-1,NARC1,PC9
Catalog No. 501659140
Encompass_Preferred
Recombinant Protein;Biotinylated Mouse PCSK9, Avi Tag (Avitag™);25UG;MABSol Biotinylated Mouse PCSK9 (PC9-M82E1) is expressed from human HEK293 cells. It contains AA Gln 35 - Gln 694 (Accession # AAH38085.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 74.9 kDa. The protein migrates as 20 kDa and 65 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation and proteolytic digestion.;Immobilized Mouse LDL R, His Tag (Cat. No. LDR-M52H8) at 10 g/mL (100 L/well) can bind Biotinylated Mouse PCSK9, His Tag (Cat. No. PC9-M82E1) with a linear range of 2-16 ng/mL (QC tested).
Catalog No. 50-165-9140 Supplier ACROBiosystems Supplier No. PC9M82E125UG
Recombinant Protein;Biotinylated Mouse PCSK9, Avi Tag (Avitag™);25UG;MABSol Biotinylated Mouse PCSK9 (PC9-M82E1) is expressed from human HEK293 cells. It contains AA Gln 35 - Gln 694 (Accession # AAH38085.1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 74.9 kDa. The protein migrates as 20 kDa and 65 kDa on a SDS-PAGE gel under reducing (R) condition due to glycosylation and proteolytic digestion.;Immobilized Mouse LDL R, His Tag (Cat. No. LDR-M52H8) at 10 g/mL (100 L/well) can bind Biotinylated Mouse PCSK9, His Tag (Cat. No. PC9-M82E1) with a linear range of 2-16 ng/mL (QC tested).